3,5-Dihydroxy-[1,2,4]triazine-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

APOSOR346340-1GEA 168 EUR
APOSOR346340-1G
3,5-Dihydroxy-[1,2,4]triazine-6-carboxylic acid
3,5-Dihydroxy-[1,2,4]triazine-6-carboxylic acid
Formel: C₄H₃N₃O₄
Molekulargewicht: 157,084 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20542230
CAS-Nummer: 13924-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR