3-Chloro-6-phenyl-pyridazine-4-carbonitrile

APOSOR346374-1GEA 0 EUR
APOSOR346374-1G
3-Chloro-6-phenyl-pyridazine-4-carbonitrile
3-Chloro-6-phenyl-pyridazine-4-carbonitrile
Formel: C₁₁H₆ClN₃
Molekulargewicht: 215,638 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00974782
CAS-Nummer: 94011-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR