3-(Chloromethyl)-5-(4-chlorophenyl)-[1,2,4]oxadiazole 95+%

Lieferant: Apollo Scientific

APOSOR346376-1GEA 265 EUR
APOSOR346376-1G
3-(Chloromethyl)-5-(4-chlorophenyl)-[1,2,4]oxadiazole 95+%
3-(Chloromethyl)-5-(4-chlorophenyl)-[1,2,4]oxadiazole
Formel: C₉H₆Cl₂N₂O
Molekulargewicht: 229,063 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01809086
CAS-Nummer: 73217-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR