2-Methylbenzo[b]thiophene-3-carboxaldehyde 95+%

Lieferant: Apollo Scientific

APOSOR346409-1GEA 150 EUR
APOSOR346409-1G
2-Methylbenzo[b]thiophene-3-carboxaldehyde 95+%
2-Methylbenzo[b]thiophene-3-carboxaldehyde
Formel: C₁₀H₈OS
Molekulargewicht: 176,235 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01651753
CAS-Nummer: 30446-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR