5-(3-Chlorophenyl)-3-methylisoxazole-4-carboxaldehyde 95+%

Lieferant: Apollo Scientific

APOSOR346423-1GEA 286 EUR
APOSOR346423-1G
5-(3-Chlorophenyl)-3-methylisoxazole-4-carboxaldehyde 95+%
5-(3-Chlorophenyl)-3-methylisoxazole-4-carboxaldehyde
Formel: C₁₁H₈ClNO₂
Molekulargewicht: 221,64 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD15142856
CAS-Nummer: 1208081-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR