3-Methylisothiazole-5-carboxaldehyde 95+%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR346460-500MGEA
449
EUR
APOSOR346460-500MG
APOSOR346460-1G
3-Methylisothiazole-5-carboxaldehyde 95+%
3-Methylisothiazole-5-carboxaldehyde
Formel:
C₅H₅NOS Molekulargewicht: 127,164 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD10700240 CAS-Nummer: 88511-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...