6-Methoxybenzo[b]thiophene-2-carboxaldehyde 95+%

Lieferant: Apollo Scientific

APOSOR346468-1GEA 308 EUR
APOSOR346468-1G
6-Methoxybenzo[b]thiophene-2-carboxaldehyde 95+%
6-Methoxybenzo[b]thiophene-2-carboxaldehyde
Formel: C₁₀H₈O₂S
Molekulargewicht: 192,234 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11101038
CAS-Nummer: 1001203-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR