2-Methyl-1,3-benzoxazole-6-carbonyl chloride 95+%

Lieferant: Apollo Scientific

APOSOR346517-1GEA 412 EUR
APOSOR346517-1G
2-Methyl-1,3-benzoxazole-6-carbonyl chloride 95+%
2-Methyl-1,3-benzoxazole-6-carbonyl chloride
Formel: C₉H₆ClNO₂
Molekulargewicht: 195,602 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13174295
CAS-Nummer: 202195-57-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR