2-(4-Methoxyphenyl)-4,5-dimethyl-1,3-thiazole 95+%

Lieferant: Apollo Scientific

APOSOR346574-1GEA 371 EUR
APOSOR346574-1G
2-(4-Methoxyphenyl)-4,5-dimethyl-1,3-thiazole 95+%
2-(4-Methoxyphenyl)-4,5-dimethyl-1,3-thiazole
Formel: C₁₂H₁₃NOS
Molekulargewicht: 219,303 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16310719
CAS-Nummer: 100192-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR