1-(3,7-Dimethyl-quinoxalin-2-yl)ethanone 95+%

Lieferant: Apollo Scientific

APOSOR346583-1GEA 622 EUR
APOSOR346583-1G
1-(3,7-Dimethyl-quinoxalin-2-yl)ethanone 95+%
1-(3,7-Dimethyl-quinoxalin-2-yl)ethanone
Formel: C₁₂H₁₂N₂O
Molekulargewicht: 200,236 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD30725949
CAS-Nummer: 38953-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR