2-Bromo-1-(3-methyl-quinoxalin-2-yl)ethanone 95+%

Lieferant: Apollo Scientific

APOSOR346584-1GEA 518 EUR
APOSOR346584-1G
2-Bromo-1-(3-methyl-quinoxalin-2-yl)ethanone 95+%
2-Bromo-1-(3-methyl-quinoxalin-2-yl)ethanone
Formel: C₁₁H₉BrN₂O
Molekulargewicht: 265,106 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05662523
CAS-Nummer: 5498-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR