5-Ethyl-2-methyl-2H-pyrazol-3-ol 95+%

Lieferant: Apollo Scientific

APOSOR346602-1GEA 371 EUR
APOSOR346602-1G
5-Ethyl-2-methyl-2H-pyrazol-3-ol 95+%
5-Ethyl-2-methyl-2H-pyrazol-3-ol
Formel: C₆H₁₀N₂O
Molekulargewicht: 126,156 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD21666882
CAS-Nummer: 29939-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR