5-Chloro-3-isopropyl-1-methyl-1H-pyrazole 95+%
Lieferant: Apollo Scientific
APOSOR346603-1GEA
518
EUR
APOSOR346603-1G
5-Chloro-3-isopropyl-1-methyl-1H-pyrazole 95+%
5-Chloro-3-isopropyl-1-methyl-1H-pyrazole
Formel:
C₇H₁₁ClN₂ Molekulargewicht: 158,629 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05860084 CAS-Nummer: 29938-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...