1-[4-(Pyridin-2-yl)phenyl]ethanone 95+%

Lieferant: Apollo Scientific

APOSOR346610-1GEA 622 EUR
APOSOR346610-1G
1-[4-(Pyridin-2-yl)phenyl]ethanone 95+%
1-[4-(Pyridin-2-yl)phenyl]ethanone
Formel: C₁₃H₁₁NO
Molekulargewicht: 197,233 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11932624
CAS-Nummer: 173681-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR