3-(Pyridin-3-yl)acrylic acid methyl ester 95+%

Lieferant: Apollo Scientific

APOSOR346615-1GEA 328 EUR
APOSOR346615-1G
3-(Pyridin-3-yl)acrylic acid methyl ester 95+%
3-(Pyridin-3-yl)acrylic acid methyl ester
Formel: C₉H₉NO₂
Molekulargewicht: 163,173 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00970044
CAS-Nummer: 61859-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR