3-Methylfuran-2-carboxamide 95+%

Lieferant: Apollo Scientific

APOSOR346630-1GEA 308 EUR
APOSOR346630-1G
3-Methylfuran-2-carboxamide 95+%
3-Methylfuran-2-carboxamide
Formel: C₆H₇NO₂
Molekulargewicht: 125,125 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11643939
CAS-Nummer: 84374-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR