5-tert-Butyl-3-cyclopropyl-1,2,4-oxadiazole 95+%

Lieferant: Apollo Scientific

APOSOR346655-1GEA 518 EUR
APOSOR346655-1G
5-tert-Butyl-3-cyclopropyl-1,2,4-oxadiazole 95+%
5-tert-Butyl-3-cyclopropyl-1,2,4-oxadiazole
Formel: C₉H₁₄N₂O
Molekulargewicht: 166,22 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD29968177
CAS-Nummer: 1165931-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR