3-Methylbenzofuran-5-carboxylic acid 95+%

Lieferant: Apollo Scientific

APOSOR346680-1GEA 729 EUR
APOSOR346680-1G
3-Methylbenzofuran-5-carboxylic acid 95+%
3-Methylbenzofuran-5-carboxylic acid
Formel: C₁₀H₈O₃
Molekulargewicht: 176,169 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07437982
CAS-Nummer: 501892-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR