5-tert-Butyl-2-methylfuran-3-carboxamide 95+%

Lieferant: Apollo Scientific

APOSOR346683-1GEA 622 EUR
APOSOR346683-1G
5-tert-Butyl-2-methylfuran-3-carboxamide 95+%
5-tert-Butyl-2-methylfuran-3-carboxamide
Formel: C₁₀H₁₅NO₂
Molekulargewicht: 181,232 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00276989
CAS-Nummer: 175276-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR