4-Bromo-3-chlorobenzamide 95+%

Lieferant: Apollo Scientific

APOSOR346690-1GEA 251 EUR
APOSOR346690-1G APOSOR346690-5G
4-Bromo-3-chlorobenzamide 95+%
4-Bromo-3-chlorobenzamide
Formel: C₇H₅BrClNO
Molekulargewicht: 234,478 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD17779957
CAS-Nummer: 1228826-41-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR