4-[tert-Butyl(4-methoxybenzyl)sulphamoyl]benzeneboronic acid 98%

APOSOR3494-1GEA 0 EUR
APOSOR3494-1G
4-[tert-Butyl(4-methoxybenzyl)sulphamoyl]benzeneboronic acid 98%
4-[tert-Butyl(4-methoxybenzyl)sulphamoyl]benzeneboronic acid
Formel: C₁₈H₂₄BNO₅S
Molekulargewicht: 377,263 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09027251
CAS-Nummer: 957060-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR