6-Bromo-5-methoxy-1H-indole

APOSOR350035-250MGEA 0 EUR
APOSOR350035-250MG
6-Bromo-5-methoxy-1H-indole
6-Bromo-5-methoxy-1H-indole
Formel: C₉H₈BrNO
Molekulargewicht: 226,07 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11848542
CAS-Nummer: 106103-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR