2-(3-Chlorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Lieferant: Apollo Scientific

APOSOR350108-1GEA 172 EUR
APOSOR350108-1G APOSOR350108-250MG
2-(3-Chlorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(3-Chlorobenzyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₁₃H₁₈BClO₂
Molekulargewicht: 252,545 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10698516
CAS-Nummer: 517920-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR