D-Xylopyranose

Lieferant: Apollo Scientific

APOSOR350116-100GEA 66.9 EUR
APOSOR350116-100G
D-Xylopyranose
D-Xylopyranose
Formel: C₅H₁₀O₅
Molekulargewicht: 150,13 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00064360
CAS-Nummer: 6763-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR