1H-Indazole-3-carboxaldehyde hydrochloride

APOSOR350191-250MGEA 0 EUR
APOSOR350191-250MG
1H-Indazole-3-carboxaldehyde hydrochloride
1H-Indazole-3-carboxaldehyde hydrochloride
Formel: C₈H₆N₂O·ClH
Molekulargewicht: 182,607 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11656791
CAS-Nummer: 1186663-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR