2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)acetic acid

Lieferant: Apollo Scientific

APOSOR350194-1GEA 224 EUR
APOSOR350194-1G
2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)acetic acid
2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)acetic acid
Formel: C₉H₈N₂O₂
Molekulargewicht: 176,172 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10698203
CAS-Nummer: 1912-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR