5-Nitro-1,2,3-benzenetricarboxylic Acid

Lieferant: Apollo Scientific

Achtung

APOSOR350301-5GEA 181 EUR
APOSOR350301-5G
5-Nitro-1,2,3-benzenetricarboxylic Acid
5-Nitro-1,2,3-benzenetricarboxylic Acid
Formel: C₉H₅NO₈
Molekulargewicht: 255,138 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03839939
CAS-Nummer: 3807-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR