2-(3-Bromophenyl)-N-tert-butylacetamide 98%
APOSOR3539-5GEA
0
EUR
APOSOR3539-5G
APOSOR3539-1G
2-(3-Bromophenyl)-N-tert-butylacetamide 98%
2-(3-Bromophenyl)-N-tert-butylacetamide
Formel:
C₁₂H₁₆BrNO Molekulargewicht: 270,166 g/mol Schmelzpunkt: 104…107 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD02666610 CAS-Nummer: 883801-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...