4-Julolidine boronic acid 95%

Lieferant: Apollo Scientific

Achtung

APOSOR360246-5GEA 1180 EUR
APOSOR360246-5G APOSOR360246-1G
4-Julolidine boronic acid 95%
4-Julolidine boronic acid
Formel: C₁₂H₁₆BNO₂
Molekulargewicht: 217,072 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03001757
CAS-Nummer: 391248-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR