5-Carboxyfuran-2-boronic acid 96%
Lieferant: Apollo Scientific
APOSOR360876-250MGEA
109
EUR
APOSOR360876-250MG
5-Carboxyfuran-2-boronic acid 96%
5-Carboxyfuran-2-boronic acid
Formel:
C₅H₅BO₅ Molekulargewicht: 155,901 g/mol Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD04113807 CAS-Nummer: 852228-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...