5-Carboxy-2-methylphenylboronic acid 97%
Lieferant: Apollo Scientific
APOSOR360884-250MGEA
108
EUR
APOSOR360884-250MG
5-Carboxy-2-methylphenylboronic acid 97%
5-Carboxy-2-methylphenylboronic acid
Formel:
C₈H₉BO₄ Molekulargewicht: 179,966 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08061639 CAS-Nummer: 170230-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...