4,5-Dihydrofuran-3-boronic acid, pinacol ester

Lieferant: Apollo Scientific

Achtung

APOSOR361128-1GEA 1280 EUR
APOSOR361128-1G APOSOR361128-250MG
4,5-Dihydrofuran-3-boronic acid, pinacol ester
4,5-Dihydrofuran-3-boronic acid, pinacol ester
Formel: C₁₀H₁₇BO₃
Molekulargewicht: 196,051 g/mol
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD13182120
CAS-Nummer: 1046812-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR