2-(Methoxycarbonyl)ethylboronic acid, pinacol ester

Lieferant: Apollo Scientific

Achtung

APOSOR361330-5GEA 183 EUR
APOSOR361330-5G APOSOR361330-25G
2-(Methoxycarbonyl)ethylboronic acid, pinacol ester
2-(Methoxycarbonyl)ethylboronic acid, pinacol ester
Formel: C₁₀H₁₉BO₄
Molekulargewicht: 214,066 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10700159
CAS-Nummer: 1150561-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR