4-Methoxycarbonyl-3-nitrophenylboronic acid 98%

Lieferant: Apollo Scientific

APOSOR361348-1GEA 129 EUR
APOSOR361348-1G
4-Methoxycarbonyl-3-nitrophenylboronic acid 98%
4-Methoxycarbonyl-3-nitrophenylboronic acid
Formel: C₈H₈BNO₆
Molekulargewicht: 224,963 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12032027
CAS-Nummer: 85107-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR