3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

APOSOR361643-100MGEA 0 EUR
APOSOR361643-100MG APOSOR361643-250MG
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
Formel: C₁₃H₁₇BN₂O₂
Molekulargewicht: 244,097 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07368014
CAS-Nummer: 937366-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR