(E)-3-Cyclopentylacrylaldehyde ≥98%
Lieferant: Apollo Scientific
APOSOR370016-1GEA
690
EUR
APOSOR370016-1G
(E)-3-Cyclopentylacrylaldehyde ≥98%
(E)-3-Cyclopentylacrylaldehyde
Formel:
C₈H₁₂O Molekulargewicht: 124,18 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD15528962 CAS-Nummer: 118235-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...