3-(7-Bromo-2,3-dihydrobenzofuran-5-yl)propanoic acid ≥98%

Lieferant: Apollo Scientific

APOSOR370088-1GEA 912 EUR
APOSOR370088-1G
3-(7-Bromo-2,3-dihydrobenzofuran-5-yl)propanoic acid ≥98%
3-(7-Bromo-2,3-dihydrobenzofuran-5-yl)propanoic acid
Formel: C₁₁H₁₁BrO₃
Molekulargewicht: 271,107 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD29053188
CAS-Nummer: 196597-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR