3-Bromo-5-chloro-6-ethylpyrazine-2-carbonitrile ≥98%

Lieferant: Apollo Scientific

APOSOR370091-1GEA 75.7 EUR
APOSOR370091-1G
3-Bromo-5-chloro-6-ethylpyrazine-2-carbonitrile ≥98%
3-Bromo-5-chloro-6-ethylpyrazine-2-carbonitrile
Formel: C₇H₅BrClN₃
Molekulargewicht: 246,492 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD30532902
CAS-Nummer: 2043020-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR