3-Methyl-1-(oxetan-3-yl)piperazine 95%

APOSOR370104-1GEA 0 EUR
APOSOR370104-1G
3-Methyl-1-(oxetan-3-yl)piperazine 95%
3-Methyl-1-(oxetan-3-yl)piperazine
Formel: C₈H₁₆N₂O
Molekulargewicht: 156,225 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD21848905
CAS-Nummer: 2070014-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR