2-Methyl-1H-indole-4-carboxaldehyde 96%

APOSOR370132-1GEA 0 EUR
APOSOR370132-1G
2-Methyl-1H-indole-4-carboxaldehyde 96%
2-Methyl-1H-indole-4-carboxaldehyde
Formel: C₁₀H₉NO
Molekulargewicht: 159,185 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12923969
CAS-Nummer: 321922-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR