6-Benzyl-1-oxa-6-azaspiro[2.5]octane 95%
Lieferant: Apollo Scientific
APOSOR370134-1GEA
724
EUR
APOSOR370134-1G
6-Benzyl-1-oxa-6-azaspiro[2.5]octane 95%
6-Benzyl-1-oxa-6-azaspiro[2.5]octane
Formel:
C₁₃H₁₇NO Molekulargewicht: 203,28 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06808592 CAS-Nummer: 19867-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...