Ethyl 3-(fur-3-yl)-3-oxopropanoate 95+%

APOSOR400173-250MGEA 0 EUR
APOSOR400173-250MG
Ethyl 3-(fur-3-yl)-3-oxopropanoate 95+%
Ethyl 3-(fur-3-yl)-3-oxopropanoate
Formel: C₉H₁₀O₄
Molekulargewicht: 182,173 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011566
CAS-Nummer: 36878-91-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR