Methyl 4-cyano-2-methoxybenzoate 95+%

APOSOR400406-5GEA 0 EUR
APOSOR400406-5G APOSOR400406-10G APOSOR400406-1G
Methyl 4-cyano-2-methoxybenzoate 95+%
Methyl 4-cyano-2-methoxybenzoate
Formel: C₁₀H₉NO₃
Molekulargewicht: 191,183 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18399095
CAS-Nummer: 406719-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR