4-Bromo-5-methoxybenzene-1,2-diamine 95+%

APOSOR400541-1GEA 0 EUR
APOSOR400541-1G
4-Bromo-5-methoxybenzene-1,2-diamine 95+%
4-Bromo-5-methoxybenzene-1,2-diamine
Formel: C₇H₉BrN₂O
Molekulargewicht: 217,063 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD21648471
CAS-Nummer: 108447-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR