4-Amino-6-bromo-7-methylquinoline

Lieferant: Apollo Scientific

APOSOR42035-1GEA 286 EUR
APOSOR42035-1G
4-Amino-6-bromo-7-methylquinoline
4-Amino-6-bromo-7-methylquinoline
Formel: C₁₀H₉BrN₂
Molekulargewicht: 237,096 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12674994
CAS-Nummer: 1189106-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR