N-Isopropyl-4-(piperidin-1-yl)benzylamine
Lieferant: Apollo Scientific
APOSOR4372-1GEA
210
EUR
APOSOR4372-1G
N-Isopropyl-4-(piperidin-1-yl)benzylamine
N-Isopropyl-4-(piperidin-1-yl)benzylamine
Formel:
C₁₅H₂₄N₂ Molekulargewicht: 232,364 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07368526 CAS-Nummer: 852180-57-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...