[4-(1H-Imidazol-2-yl)phenyl]boronic acid, pinacol ester

Lieferant: Apollo Scientific

APOSOR46750-250MGEA 391 EUR
APOSOR46750-250MG
[4-(1H-Imidazol-2-yl)phenyl]boronic acid, pinacol ester
[4-(1H-Imidazol-2-yl)phenyl]boronic acid, pinacol ester
Formel: C₁₅H₁₉BN₂O₂
Molekulargewicht: 270,135 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18383629
CAS-Nummer: 1229584-17-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR