3-(Boc-amino)-3-(4-bromophenyl)-1-propanol

Lieferant: Apollo Scientific

APOSOR470324-5GEA 1540 EUR
APOSOR470324-5G
3-(Boc-amino)-3-(4-bromophenyl)-1-propanol
3-(Boc-amino)-3-(4-bromophenyl)-1-propanol
Formel: C₁₄H₂₀BrNO₃
Molekulargewicht: 330,217 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04115556
CAS-Nummer: 898404-64-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR