4-Oxo-1-(o-tolyl)cyclohexanecarboxylic acid

Lieferant: Apollo Scientific

APOSOR470534-5GEA 1010 EUR
APOSOR470534-5G
4-Oxo-1-(o-tolyl)cyclohexanecarboxylic acid
4-Oxo-1-(o-tolyl)cyclohexanecarboxylic acid
Formel: C₁₄H₁₆O₃
Molekulargewicht: 232,275 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22423141
CAS-Nummer: 1385694-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR