(Phenyl)(3-quinolyl)methanol

Lieferant: Apollo Scientific

APOSOR470548-1GEA 499 EUR
APOSOR470548-1G
(Phenyl)(3-quinolyl)methanol
(Phenyl)(3-quinolyl)methanol
Formel: C₁₆H₁₃NO
Molekulargewicht: 235,281 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12833814
CAS-Nummer: 37045-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR